Learn More
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper
Borosilicate volumetric flask with glass stopper and USP certificate.
Brand: BRAND™ 236984
1030.56 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Borosilicate 3.3
- To DIN EN ISO 1042
- Calibrated to contain (TC, In)
- Includes USP certificate
Specifications
USP Single Batch | |
Borosilicate Glass (3.3) | |
Reusable | |
Narrow | |
Flat |
2 L | |
Autoclavable | |
Yes | |
Non-sterile |
Product Content Correction
Product Title
BRAND™ Class A Borosilicate Volumetric Flask with Glass Stopper > Volumetric Flask, 2 L, USP Single Batch Certificate
Spot an opportunity for improvement?Share a Content Correction