Learn More
Abeba™ Classic 31001 Shoes
Animal fats, oil and fuel resistant. Leather/microfiber construction.
Brand:  Abeba™ 31001/41
Additional Details : Weight : 1.02500kg
 View more versions of this product
Product Code. 15425734
  • 946.10 SEK / 1 pair
Estimated Shipment: 17-05-2024
to see stock.
Add to basket
Description
Description
  • CE EN ISO 20345:2011 SB A, E, FO SRC
  • Microfiber
  • White
  • Steel toe cap
  • Slip resistance PU outsole sole
  • ESD dissipative according to DIN EN 61340
  • Heel strap non-foldable, adjustable
  • Microfiber lining and heel patch
  • Adjustable instep area
  • Buckle fastening
  • Comfortable shape
Specifications
Specifications
CE EN ISO 20345:2011 SB A, E, FO SRC
Microfiber
41
Polyurethane
Steel
White
Microfiber
Slip-resistant
31001
Animal fats, oil and fuel