missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLCC1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26680-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Neurofibromin is a member of the GTPase-activating (GAP) class of proteins. These proteins enhance the GTPase activity of signaling G-Proteins like Ras. The NF1 gene is also classified as a tumor suppressor gene, and many studies have shown that mutations at the NF1 locus produce neurofibromas, rhabdomyosarcomas, leimyosarcomas, mental retardation and memory deficiency. Neurofibromin has also been associated with the intermediated filaments of the cytoskeleton. NF1 is expressed at its highest levels during periods of desmosome formation and mutations of NF1 produce variable cell sizes and morphologies.Specifications
CLCC1 | |
Polyclonal | |
Unconjugated | |
CLCC1 | |
chloride channel CLIC like 1, CLCC1, KIAA0761, MCLC | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23155 | |
-20°C | |
Liquid |
Western Blot | |
0.47 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96S66 | |
CLCC1 | |
CLCC1 Fusion Protein Ag24682 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |