missing translation for 'onlineSavingsMsg'
Learn More

CLDN16 Rabbit anti-Canine, Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27638-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16825755

  • 1656.73 SEK / 20µL
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Tight junction group of proteins are a cell-to-cell adhesion structure in epithelial cells that constitute the epithelial junctional complex with adherens junctions and desmosomes. Tight junction strands are mainly composed of claudins, occludin, and JAM. Claudin-16 belongs to the Claudin family of protein which is composed of 23 integral membrane proteins. Claudins are responsible for the formation of tight junction strands and are connected with the actin cytoskeleton mediated by ZO-1. Claudin 16, also known as Paracellin-1, is a renal tight junction protein required for paracellular magnesium resorption. It is localized in the intercellular junctions of the epithelial cells of the thick ascending limb of Henle′s loop (a TAL marker) are used to localize this specific structure of nephrons in kidney sections. Mg2+ resorption occurs in TAL, and Claudin-16 is required for this paracellular function. Failure in renal ′handling′ of magnesium can result in hypermagnesemia, whose major direct toxicity implications are cardiovascular as well as renal failure. Recent studies have shown that the lack of Claudin-16 proteins contributes to the dysfunction of paracellular renal transport systems.
Specifications

Specifications

CLDN16
Polyclonal
Unconjugated
CLDN16
claudin 16, CLDN16, HOMG3, Paracellin 1, PCLN 1, PCLN1
Rabbit
Antigen Affinity Chromatography
RUO
10686, 608218
-20°C
Liquid
Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y5I7
CLDN16
CLDN16 Fusion Protein Ag26531
20 μL
Primary
Human, Canine
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers