Learn More
CLEC3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20311-1-AP-150UL
 View more versions of this product
Product Code. 16889834
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
Specifications
Specifications
CLEC3B
Polyclonal
Unconjugated
CLEC3B
CLEC3B, Tetranectin, TN, TNA
Rabbit
Antigen Affinity Chromatography
RUO
21922, 7123
-20°C
Liquid
Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05452, P43025
CLEC3B
CLEC3B Fusion Protein Ag14147
150 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CLEC3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated