Learn More
CLEC3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20311-1-AP-150UL
 View more versions of this product
Product Code. 16889834
  • 4652.90 SEK / 150µL
Description
Tetranectin (TN) is a serum and tissue protein, a C-type lectin, which binds to Ca++. It is a homotrimer of monomers each with a mass of 20 kDa, plasma or serum concentrations of TN are found to be approximately 10 mg/l. In vitro, TN can bind to kringle 4 of plasminogen and enhance the activation of plaminogen to plasmin, catalyzed by tissue plasminogen activator in the presence of poly-D-lysine. TN is best known as a prognostic marker in ovarian cancer.
Specifications
Specifications
CLEC3B
Polyclonal
Unconjugated
CLEC3B
CLEC3B, Tetranectin, TN, TNA
Rabbit
Antigen Affinity Chromatography
RUO
21922, 7123
-20°C
Liquid
Immunoprecipitation, Western Blot
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P05452, P43025
CLEC3B
CLEC3B Fusion Protein Ag14147
150 μL
Primary
Human, Mouse
Antibody
IgG
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CLEC3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated