missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLIC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12298-2-AP-20UL
This item is not returnable.
View return policy
Description
Chloride channels are a diverse group of proteins that regulate fundamental cellular processes including stabilization of cell membrane potential, transepithelial transport, maintece of intracellular pH, and regulation of cell volume. Chloride intracellular channel 4 (CLIC4) protein, encoded by the CLIC4 gene, is a member of the p64 family; the gene is expressed in many tissues and exhibits a intracellular vesicular pattern in Panc-1 cells (pancreatic cancer cells).Specifications
CLIC4 | |
Polyclonal | |
Unconjugated | |
CLIC4 | |
CLIC4, CLIC4L, H1, huH1, MTCLIC, p64H1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
25932, 29876, 83718 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QYB1, Q9Y696, Q9Z0W7 | |
Clic4 | |
CLIC4 Fusion Protein Ag2942 | |
20 μL | |
Primary | |
Mouse, Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CLIC4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction