Learn More
Memmert™ Clip for 100 mL Flask
Clip for 100 mL flask (12 flasks max.)
Brand:  Memmert™ WTB15B02692
261.02 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17150305
  • 273.00 SEK / Each
Currently Unavailable: We have no information when this product will be available.
View alternative products
Description
Description
  • Accessory
  • Max. flasks see sketch on demand
Specifications
Specifications
WTB15
Clip for 100 mL flask
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Clip for 100 mL Flask > WTB15