Learn More
Memmert™ Clip for 100 mL Flask
Clip for 100 mL flask (12 flasks max.)
Brand: Memmert™ WTB24B02692
261.02 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- Max. flasks see sketch on demand
Specifications
WTB24 | |
Clip for 100 mL flask |
Product Content Correction
Product Title
Memmert™ Clip for 100 mL Flask > WTB24
Spot an opportunity for improvement?Share a Content Correction