Learn More
Memmert™ Clip for 300 mL Flask
Clip for 300 mL flask (14 flasks max.)
Brand: Memmert™ WTB15B02694
329.11 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Accessory
- Max. flasks see sketch on demand
Specifications
WTB15 | |
Clip for 300 mL flask |
Product Content Correction
Product Title
Memmert™ Clip for 300 mL Flask > WTB15
Spot an opportunity for improvement?Share a Content Correction