Learn More
Memmert™ Clip for 500 mL Flask
Clip for 500 mL flask (5 flasks max.)
368.00 SEK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17120395
|
Memmert™
WTB50B02695 |
WTB50 |
368.00 SEK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
|||||||||
17180305
|
Memmert™
WTB15B02695 |
WTB15 |
368.00 SEK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
|||||||||
17190335
|
Memmert™
WTB24B02695 |
WTB24 |
368.00 SEK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
|||||||||
17190365
|
Memmert™
WTB35B02695 |
WTB35 |
368.00 SEK
Each |
N/A | |||||
Currently Unavailable: We have no information when this product will be available.
View alternative products
|
Description
- Accessory
- Max. flasks see sketch on demand
Specifications
Clip for 500 mL flask |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ Clip for 500 mL Flask