Learn More
Memmert™ Clips for Flask
Brand:  Memmert™ B02694
337.81 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12018471
  • 344.00 SEK / Each
Only null left
Estimated Shipment: 24-07-2025
Specifications
14L, 22L, 29L, 45L bath
Clip
For 300mL flask; 6 flasks maximum
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Memmert™ Clips for Flask > For 300mL flask; 6 flasks maximum