missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CLK1 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
CĂłdigo de producto | Marca | Quantity | Precio | Cantidad y disponibilidad | |||||
16879974
|
Proteintech
20439-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869974
|
Proteintech
20439-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
DescripciĂłn
This gene encodes a member of the CDC2-like (or LAMMER) family of dual specificity protein kinases. In the nucleus, the encoded protein phosphorylates serine/arginine-rich proteins involved in pre-mRNA processing, releasing them into the nucleoplasm. The choice of splice sites during pre-mRNA processing may be regulated by the concentration of transacting factors, including serine/arginine rich proteins. Therefore, the encoded protein may play an indirect role in governing splice site selection.Especificaciones
CLK1 | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
P22518, P49759 | |
1195, 12747, 301434 | |
CLK1 Fusion Protein Ag14214 | |
Primary | |
-20°C | |
CLK1 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDC like kinase 1, CLK, CLK/STY, CLK1, STY | |
CLK1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
ÂżDetecta una oportunidad de mejora?Comparta una correcciĂłn de contenido
CorrecciĂłn del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
TĂtulo del producto
CLK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech