Läs mer
Cloxacillin sodium salt, Thermo Scientific™
A semisynthetic antibiotic
Brand:  Thermo Scientific Alfa Aesar J64314.03
Kvantitet 1g
 Se fler versioner av denna produkt
Produktkod. 15406319
  • 787.86 SEK / 1g
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
Beskrivning
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Cloxacillin sodium salt is used to treat infections caused by penicillin G-resistant staphylococci. It has been used in studies of topical antibacterial agent for burn and crush wounds. It is used as an AmpC β-lactamase inhibitor in combination with β-lactam antibiotics.
Solubility
Soluble in water
Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiska identifierare
642-78-4
458.869
JBJAWVZHLGPBJA-SLINCCQESA-N
(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
C19H18ClN3NaO5S
MFCD00063568
66587017
CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na]
Specifikationer
Cloxacillin sodium salt
White
C19H18ClN3NaO5S
4103180
Soluble in water
CC1=C(C(=NO1)C2=CC=CC=C2Cl)C(=O)NC3C4N(C3=O)C(C(S4)(C)C)C(=O)O.[Na]
458.869
457.86
642-78-4
1g
MFCD00063568
14,2419
JBJAWVZHLGPBJA-SLINCCQESA-N
(2S,5R,6R)-6-[[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid;sodium
66587017