missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech
Mouse Monoclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CLTCL1 |
---|---|
Clone | 1B5A10 |
Concentration | 1.2 mg/mL |
Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887995
|
Proteintech
66274-1-IG-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16877995
|
Proteintech
66274-1-IG-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The antibody is specific to CLTCL1. It has no cross reaction to CLTC.
Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.Specifications
CLTCL1 | |
1.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CHC22, Clathrin heavy chain 2, clathrin, heavy chain like 1, CLH 22, CLH22, CLTCL, CLTCL1, CLTD | |
CLTCL1 | |
IgG2a | |
Protein A | |
Antibody |
1B5A10 | |
Western Blot, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P53675 | |
8218 | |
Peptide | |
Primary | |
-20°C | |
CLTCL1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CLTCL1 Mouse anti-Human, Clone: 1B5A10, Proteintech