Läs mer
Skytec™ CMBH-panndräkt
Beskrivning
- Ring-pull zip for easy donning and doffing when wearing gloves
- Double storm-facing stud closure for increased protection
- Helmet-compatible hood
- Raglan sleeve for greater freedom of movement
- Garment can be reused if not contaminated or damaged
- Stitched and high-frequency welded seams
- Tested to ASTM F903
Specifikationer
Specifikationer
| Färg | Green |
| Huva typ | Attached |
| Material | PVC |
| Storlek | Large |
| Ankel stil | Loose |
| Förslutningstyp | Zipper |
| Manschettstil | Standard |
| Produkttyp | Chemical Protective Clothing |
| Sömtyp | Welded |