Learn More
CMPK1 Mouse anti-Human, Unconjugated, Clone: 2D11G7, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67519-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840196
  • 1656.73 SEK / 20µL
Estimated Shipment: 23-04-2024
to see stock.
Description
Description
The CMPK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CMPK1. This antibody recognizes human antigen. The CMPK1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
CMPK1
Monoclonal
500 μg/mL
Mouse
Primary
Human
Western Blot, Immunofluorescence, ELISA
2D11G7
Unconjugated
20 μL
51727
IgG2b