Learn More
CMPK1 Mouse anti-Human, Unconjugated, Clone: 2D11G7, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67519-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The CMPK1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CMPK1. This antibody recognizes human antigen. The CMPK1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
CMPK1 | |
Monoclonal | |
500 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunofluorescence, ELISA | |
2D11G7 | |
Unconjugated | |
20 μL | |
51727 | |
IgG2b |