Learn More
CNBP Mouse anti-Human, Unconjugated, Clone: 1H1D12, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67109-1-IG-150UL
 View more versions of this product
Product Code. 16879545
  • 4652.90 SEK / 150µL
Description
The CNBP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNBP. This antibody recognizes human, mouse, rat antigen. The CNBP antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
Specifications
CNBP
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, ELISA
1H1D12
Unconjugated
150 μL
7555
IgG2a
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CNBP Mouse anti-Human, Unconjugated, Clone: 1H1D12, Proteintech > 150 μL