Learn More
CNBP Mouse anti-Human, Unconjugated, Clone: 1H1D12, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67109-1-IG-150UL
Description
The CNBP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CNBP. This antibody recognizes human, mouse, rat antigen. The CNBP antibody has been validated for the following applications: IP, WB, ELISA analysis.
Specifications
CNBP | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunoprecipitation, ELISA | |
1H1D12 | |
Unconjugated | |
150 μL | |
7555 | |
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.