missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21073-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene, which can bind actin, calmodulin, troponin C, and tropomyosin, may function in the structural organization of actin filaments. The encoded protein could play a role in smooth muscle contraction and cell adhesion. Two transcript variants encoding different isoforms have been found for this gene.Specifications
CNN2 | |
Polyclonal | |
Unconjugated | |
CNN2 | |
calponin 2, Calponin H2, smooth muscle, CNN2, Neutral calponin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1265, 12798, 690976 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q08093, Q99439 | |
Cnn2 | |
CNN2 Fusion Protein Ag15225 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CNN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction