Learn More
CNNM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14066-1-AP-150UL
 View more versions of this product
Product Code. 16882534
  • 4652.90 SEK / 150µL
This item is not returnable. View return policy
Description
Description
This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.
Specifications
Specifications
CNNM4
Polyclonal
Unconjugated
CNNM4
ACDP4, CNNM4, cyclin M4, KIAA1592, Metal transporter CNNM4
Rabbit
Antigen Affinity Chromatography
RUO
26504, 363216, 94220
-20°C
Liquid
Immunoprecipitation, Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P0C588, Q69ZF7, Q6P4Q7
Cnnm4
CNNM4 Fusion Protein Ag5121
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CNNM4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated