missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNTN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13843-1-AP-150UL
This item is not returnable.
View return policy
Description
Contactin 1 encodes a protein that is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system. Two alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
CNTN1 | |
Polyclonal | |
Unconjugated | |
CNTN1 | |
CNTN1, contactin 1, F3, Glycoprotein gp135, GP135, Neural cell surface protein F3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117258, 1272, 12805 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P12960, Q12860, Q63198 | |
Cntn1 | |
CNTN1 Fusion Protein Ag4803 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CNTN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction