missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Coagulation Factor III/Tissue Factor Antibody (CL3807), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61641
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Coagulation Factor III/Tissue Factor Monoclonal specifically detects Coagulation Factor III/Tissue Factor in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifikationer
| Coagulation Factor III/Tissue Factor | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
| F3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: VKQTYLARVFSYPAGNVESTGSAGEPLYENSPEFTPYLETNLGQPTIQSFEQVGTKVNVTVEDERTLVRRNNTFLSLRDVFGKDLIYTLYYWKSSSSGKKTAKTNTNEFLIDVDKGEN | |
| 100 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot, Immunohistochemistry | |
| CL3807 | |
| Western Blot 1 ug/ml, Immunohistochemistry 1:2500 - 1:5000 | |
| CD142, CD142 antigen, Coagulation factor III, coagulation factor III (thromboplastin, tissue factor), FLJ17960, TF, TFA, Thromboplastin, tissue factor | |
| Mouse | |
| Protein A purified | |
| Cancer | |
| 2152 | |
| Human | |
| Purified |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering