Learn More
Cobalt Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical Product Code.: 10356090
Cobalt Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical
Product Code. 10356090 Shop All Fisher Chemical Products
100 mL, 100mL
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 10356090
Brand: Fisher Chemical J/8250/05
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Packaging HDPE plastic bottle
Physical Form Liquid
MDL Number 10935
pH 1
Quantity 100 mL
compliance-icons compliance-icons
Product Identifier
  • Cobalt solution 10 000 ppm
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Product Title
Cobalt Standard Metal Solution 10,000ppm, for ICP, Solution HNO3 1M, Fisher Chemical >