missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14277-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.Specifications
COG6 | |
Polyclonal | |
Unconjugated | |
COG6 | |
COD2, COG complex subunit 6, COG6, KIAA1134 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
310411, 57511, 67542 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
0.56 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FP9, Q8R3I3, Q9Y2V7 | |
COG6 | |
COG6 Fusion Protein Ag5624 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |