missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COG6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | COG6 |
---|---|
Concentration | 0.56 mg/mL |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16832854
|
Proteintech
14277-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16822854
|
Proteintech
14277-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.Specifications
COG6 | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q68FP9, Q8R3I3, Q9Y2V7 | |
310411, 57511, 67542 | |
COG6 Fusion Protein Ag5624 | |
Primary | |
-20°C | |
COG6 |
0.56 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
COD2, COG complex subunit 6, COG6, KIAA1134 | |
COG6 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |