missing translation for 'onlineSavingsMsg'
Learn More

COG6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 14277-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16832854

  • 1656.73 SEK / 20µL
Estimated Shipment: 31-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This gene encodes a subunit of the conserved oligomeric Golgi complex that is required for maintaining normal structure and activity of the Golgi apparatus. The encoded protein is organized with conserved oligomeric Golgi complex components 5, 7 and 8 into a sub-complex referred to as lobe B. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

COG6
Polyclonal
Unconjugated
COG6
COD2, COG complex subunit 6, COG6, KIAA1134
Rabbit
Antigen Affinity Chromatography
RUO
310411, 57511, 67542
-20°C
Liquid
Western Blot, Immunofluorescence, Immunocytochemistry, Flow Cytometry
0.56 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q68FP9, Q8R3I3, Q9Y2V7
COG6
COG6 Fusion Protein Ag5624
20 μL
Primary
Human, Rat, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers