Learn More
Collagen Type III (N-terminal) Rabbit anti-Human, Unconjugated, Clone: 2O1, Proteintech
Rabbit Recombinant Antibody
Brand:  Proteintech 80009-1-RR-100UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16830436
  • 5195.02 SEK / 100µL
Estimated Shipment: 15-05-2024
to see stock.
Description
Description
The Collagen Type III (N-terminal) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Collagen Type III (N-terminal). This antibody recognizes human antigen. The Collagen Type III (N-terminal) antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Specifications
Collagen Type III (N-terminal)
Monoclonal
750 μg/mL
Rabbit
Primary
Human
Immunohistochemistry, Immunofluorescence
2O1
Unconjugated
100 μL
1281