missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Collagen Type III (N-terminal) Rabbit anti-Human, Unconjugated, Clone: 2O1, Proteintech
Rabbit Recombinant Antibody
1763.23 SEK - 5195.02 SEK
Specifications
Antigen | Collagen Type III (N-terminal) |
---|---|
Clone | 2O1 |
Concentration | 750 μg/mL |
Applications | Immunohistochemistry, Immunofluorescence |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16840436
|
Proteintech
80009-1-RR-20UL |
20 μL |
1763.23 SEK
20µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16830436
|
Proteintech
80009-1-RR-100UL |
100 μL |
5195.02 SEK
100µL |
Estimated Shipment: 15-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Collagen Type III (N-terminal) antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Collagen Type III (N-terminal). This antibody recognizes human antigen. The Collagen Type III (N-terminal) antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
Collagen Type III (N-terminal) | |
750 μg/mL | |
Monoclonal | |
Rabbit | |
1281 | |
Primary |
2O1 | |
Immunohistochemistry, Immunofluorescence | |
Unconjugated | |
Human | |
IgG |