missing translation for 'onlineSavingsMsg'
Learn More

Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height, with Boxes

Comfort Rack designed to ensure a more ergonomical operating process when working with samples.

Brand:  Tenak™ TE24312B

Additional Details : Weight : 4.66000kg

 View more versions of this product

Product Code. 15270784

  • 3450.00 SEK / Each
Estimated Shipment: 01-08-2024
to see stock.
Add to basket

Description

Description

  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
  • Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications

Specifications

100 mm (Box)
Upright Freezers
3 x 5
683 x 140 x 315 mm
Boxes (15)
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers