Learn More
Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height, with Boxes
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
2375.00 SEK - 3450.00 SEK
Specifications
Height (Metric) | 100 mm (Box) |
---|---|
For Use With (Equipment) | Upright Freezers |
Material | Stainless Steel |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15210784
|
Tenak™
TE24302B |
Boxes (8) |
2625.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15230784
|
Tenak™
TE24304B |
Boxes (12) |
3020.00 SEK
Each |
Estimated Shipment: 22-05-2024
Log in to see stock. |
|||||
15250784
|
Tenak™
TE24310B |
Boxes (10) |
2985.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15270774
|
Tenak™
TE24294B |
Boxes (6) |
2375.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15270784
|
Tenak™
TE24312B |
Boxes (15) |
3450.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
15290774
|
Tenak™
TE24296B |
Boxes (9) |
2690.00 SEK
Each |
Estimated Shipment: 19-07-2024
Log in to see stock. |
|||||
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
- Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications
100 mm (Box) | |
Upright Freezers | |
Stainless Steel |