Learn More
SI Analytics™ Complete Dosing Unit
SI Analytics™ Complete Dosing Unit comes with piston and piston rod, as well as a UV protection coat for use with the TITRONIC™ Series.
Brand:  SI Analytics™ 285223767
 View more versions of this product
Product Code. 10499821
  • 4540.00 SEK / Each
Only null left
In Stock
Description
  • Dosing unit
  • Includes piston, piston rod, and UV protection coat
  • For use wtih TITRONIC™ Universal and TITRONIC™ 300
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complete Dosing Unit
Piston, Piston Rod and UV Protection Coat
50 mL
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SI Analytics™ Complete Dosing Unit > Capacity: 50mL