Learn More
Lenz Laborglasinstrumente™ Condensers with Core and Sleeve
592.00 SEK - 675.00 SEK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10403174
|
Lenz Laborglasinstrumente™
05210412 |
400 mm |
675.00 SEK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
10020944
|
Lenz Laborglasinstrumente™
05210312 |
250 mm |
592.00 SEK
Each |
Estimated Shipment: 25-07-2025
Log in to see stock. |
||||||
Specifications
Condenser | |
1 Each |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Condensers with Core and Sleeve