Learn More
SCAT Connectors for HPLC
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
Brand: SCAT 107807
95.53 SEK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
Description
SCAT™ connectors are available in the following versions:
- Conical connectors
- Y-connectors
- Tube connectors
- Capillary connectors
Specifications
11 mm | |
Tubing | |
Y-connector |
Polypropylene | |
HPLC/UHPLC Systems |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now