missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPG2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 16111-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Description
The coatomer is a cytosolic protein complex that binds to dilysine motifs and reversibly associates with Golgi non-clathrin-coated vesicles, which further mediate biosynthetic protein transport from the ER, via the Golgi up to the trans Golgi network.Specifications
COPG2 | |
Polyclonal | |
Unconjugated | |
COPG2 | |
2 COP, Coatomer subunit gamma 2, COPG2, Gamma 2 coat protein, Gamma 2 COP | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26958, 54160 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9QXK3, Q9UBF2 | |
COPG2 | |
COPG2 Fusion Protein Ag9034 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COPG2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction