Learn More
COQ6 Mouse anti-Human, Unconjugated, Clone: 2F5B1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67162-1-IG-20UL
 View more versions of this product
Product Code. 16889635
  • 1645.54 SEK / 20µL
Description
The COQ6 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human COQ6. This antibody recognizes human, mouse, rat, pig antigen. The COQ6 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
COQ6
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, ELISA
2F5B1
Unconjugated
20 μL
51004
IgG1
Certificates
Promotions
Promotions
Product Content Correction
Product Title
COQ6 Mouse anti-Human, Unconjugated, Clone: 2F5B1, Proteintech > 20 μL