Learn More
Corning™ Universal Fit Pipet Tips: Bulk Packed
Designed to assure a reliable fit with major brand pipetters
Brand: Corning™ 4862
427.54 SEK valid until 2025-08-24
Use promo code "24117" to get your promotional price.
Description
Designed to assure a reliable fit with major brand pipetters
- Beveled orifice helps ensure accurate fluid delivery
- 1–200μL tips are graduated at 10, 50 and 100μL
- 100–1000μL tips are graduated at 250, 500, and 1000μL
- Choice of sterile or nonsterile
- Bulk packed tips are nonsterile and very economical
Specifications
1 to 200 μL | |
Natural | |
Beveled, Round | |
Universal Fit | |
Polypropylene |
Non-sterile | |
Bulk | |
1000 Tips/Bag, 1000 Tips/Cs. | |
Yes |
Promotions
-
Multiple Manufacturers
Genomics Powerhouse
Power your pipeline from PCR to sequencing.Promo Code: 24117Expires: 2025-08-24
New Offer Save Up to 92%