Learn More
CORO2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13802-1-AP-150UL
 View more versions of this product
Product Code. 16882194
  • 4652.90 SEK / 150µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
May play a role in the reorganization of neuronal actin structure.
Specifications
Specifications
CORO2B
Polyclonal
Unconjugated
CORO2B
Clipin C, CLIPINC, CORO2B, Coronin 2B, Coronin 5, Coronin like protein C, KIAA0925, Protein FC96
Rabbit
Antigen Affinity Chromatography
RUO
10391, 235431, 300768
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8BH44, Q9UQ03
Coro2b
CORO2B Fusion Protein Ag4793
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CORO2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated