missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX4NB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | COX4NB |
---|---|
Concentration | 0.3 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16879354
|
Proteintech
19889-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16869354
|
Proteintech
19889-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Expressed in liver, pancreas, heart, lung, kidney, brain, skeletal muscle, and placenta, Expression levels are highest in pancreas and moderate in heart, skeletal muscle, and placenta.Specifications
COX4NB | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
O43402, O70378, Q5FVL2 | |
10328, 18117, 361425 | |
COX4NB Fusion Protein Ag13720 | |
Primary | |
-20°C | |
EMC8 |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C16orf2, C16orf4, COX4 neighbor, COX4AL, COX4NB, EMC8, FAM158B, Neighbor of COX4, NOC4, Protein FAM158B | |
Emc8 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COX4NB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech