missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX6B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11425-1-AP-20UL
This item is not returnable.
View return policy
Description
Connects the two COX monomers into the physiological dimeric form.Specifications
COX6B1 | |
Polyclonal | |
Unconjugated | |
COX6B1 | |
COX VIb 1, COX6B, COX6B1, COXG | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
110323, 1340, 688869 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14854, P56391, P80430 | |
Cox6b1 | |
COX6B1 Fusion Protein Ag1984 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COX6B1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction