missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COX7A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18122-1-AP-150UL
This item is not returnable.
View return policy
Description
This protein is one of the nuclear-coded polypeptide chains of cytochrome c oxidase, the terminal oxidase in mitochondrial electron transport.Specifications
COX7A2 | |
Polyclonal | |
Unconjugated | |
COX7A2 | |
COX7A2, COX7AL, COX7AL1, COXVIIa L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12866, 1347, 29507 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P14406, P35171, P48771 | |
Cox7a2 | |
COX7A2 Fusion Protein Ag12668 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
COX7A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction