missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CP110 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CP110 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16890674
|
Proteintech
12780-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16880674
|
Proteintech
12780-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GSG2 is required for normal alignment of chromosomes at metaphase.Specifications
CP110 | |
Immunofluorescence, Immunoprecipitation, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Canine, Human | |
O43303, Q7TSH4 | |
101565, 361634, 479826, 9738 | |
CP110 Fusion Protein Ag3489 | |
Primary | |
-20°C | |
CCP110 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Centrosomal protein of 110 kDa, CP110, CP110 protein, KIAA0419 | |
Ccp110 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CP110 Rabbit anti-Canine, Human, Mouse, Rat, Polyclonal, Proteintech