missing translation for 'onlineSavingsMsg'
Learn More

CPLX4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 21222-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16880845

  • 3665.00 SEK / 150µL
Estimated Shipment: 31-03-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

CPLX4
Polyclonal
Unconjugated
CPLX4
complexin 4, Complexin IV, CPLX4, CPX IV, CPXIV
Rabbit
Antigen Affinity Chromatography
RUO
225644, 339302, 361342
-20°C
Liquid
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.59 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z7G2, Q80WM3
CPLX4
CPLX4 Fusion Protein Ag15661
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers