missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CPT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1645.54 SEK - 4652.90 SEK
Specifications
Antigen | CPT1B |
---|---|
Concentration | 0.33 mg/mL |
Applications | Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16891705
|
Proteintech
22170-1-AP-20UL |
20 ÎĽL |
1645.54 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16881705
|
Proteintech
22170-1-AP-150UL |
150 ÎĽL |
4652.90 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene, a member of the carnitine/choline acetyltransferase family, is the rate-controlling enzyme of the long-chain fatty acid beta-oxidation pathway in muscle mitochondria. This enzyme is required for the net transport of long-chain fatty acyl-CoAs from the cytoplasm into the mitochondria. Multiple transcript variants encoding different isoforms have been found for this gene, and one of the transcripts is bicistronic.Specifications
CPT1B | |
Immunoprecipitation, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q63704, Q924X2, Q92523 | |
12895, 1375, 25756 | |
CPT1B-specific Fusion Protein Ag17840 | |
Primary | |
-20°C | |
CPT1B |
0.33 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
CPT I, CPT1 M, CPT1B, CPTI M, KIAA1670, M CPT1, MCCPT1 | |
CPT1B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CPT1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech