missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRABP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12588-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a specific binding protein for a vitamin A family member and is thought to play an important role in retinoic acid-mediated differentiation and proliferation processes. It is structurally similar to the cellular retinol-binding proteins, but binds only retinoic acid at specific sites within the nucleus, which may contribute to vitamin A-directed differentiation in epithelial tissue.Specifications
CRABP1 | |
Polyclonal | |
Unconjugated | |
CRABP1 | |
CRABP, CRABP I, CRABP1, CRABPI, RBP5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12903, 1381, 25061 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29762, P62965, P62966 | |
CRABP1 | |
CRABP1 Fusion Protein Ag3291 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CRABP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 150 μL; Unconjugated
Spot an opportunity for improvement?Share a Content Correction