Learn More
CREB3L1 OASIS Mouse anti-Human, Unconjugated, Clone: 2B5B4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67617-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840346
  • 1236.20 SEK / 20µL
Estimated Shipment: 05-10-2022
to see stock.
Description
Description
The CREB3L1, OASIS antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CREB3L1, OASIS. This antibody recognizes human, mouse, rat antigen. The CREB3L1, OASIS antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
CREB3L1 OASIS
Monoclonal
1000 μg/mL
Mouse
Primary
Mouse, Rat, Human
ELISA, Western Blot
2B5B4
Unconjugated
20 μL
90993
IgG1