Learn More
Abnova™ CREB3L1 Split FISH Probe Product Code.: 16043485
Abnova™ CREB3L1 Split FISH Probe
Product Code. 16043485
100 μL, 100µL
Click to view available options
Quantity:
100 μL
200 μL
Unit Size:
100µL
200µL
Product Code. 16043485
Brand: Abnova™ FS0049.100uL
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
Specifications
Content And Storage Store at 4°C in the dark
Format Tube
For Use With (Application) Fluorescent In Situ Hybridization (Cell)
Gene cAMP responsive element binding protein 3-like 1
Gene Alias OASIS
Gene Symbol CREB3L1
Gene ID (Entrez) 90993
Product Type DNA/RNA
Quantity 100 μL
Form Liquid
Product Content Correction
Product Title
Abnova™ CREB3L1 Split FISH Probe >