Learn More
CREB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14196-1-AP-20UL
 View more versions of this product
Product Code. 16892724
  • 1645.54 SEK / 20µL
This item is not returnable. View return policy
Description
Description
The product of this gene belongs to the CRE (cAMP response element)-binding protein family. Members of this family contain zinc-finger and bZIP DNA-binding domains. The encoded protein specifically binds to CRE as a homodimer or a heterodimer with c-Jun or CRE-BP1, and functions as a CRE-dependent trans-activator. Alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
Specifications
CREB5
Polyclonal
Unconjugated
CREB5
CRE BPA, CREB 5, CREB5, CREBPA
Rabbit
Antigen Affinity Chromatography
RUO
231991, 500131, 9586
-20°C
Liquid
Western Blot
0.39 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q02930, Q8K1L0
Creb5
CREB5 Fusion Protein Ag5399
20 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CREB5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated