Learn More
CRIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14801-1-AP-20UL
 View more versions of this product
Product Code. 16883624
  • 1645.54 SEK / 20µL
Description
CRIP2 is a 208-amino acid protein containing 2 LIM domains and shares 93% amino acid sequence identity with its rat homolog, called Esp1 or Crp2. Northern blot analysis showed widespread tissue expression of the 1.3-kb CRIP2 mRNA, with the highest level of expression found in heart. In testis, a second 1.7-kb mRNA was also detected.
Specifications
Specifications
CRIP2
Polyclonal
Unconjugated
CRIP2
CRIP, CRIP2, CRP 2, CRP2, cysteine rich protein 2, ESP1, Protein ESP1
Rabbit
Antigen Affinity Chromatography
RUO
1397, 338401, 68337
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P36201, P52943, Q9DCT8
Crip2
CRIP2 Fusion Protein Ag6515
20 μL
Primary
Rat, Human, Mouse
Antibody
IgG
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
CRIP2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated