missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15724-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The function of this protein remains unknown.Specifications
CRTAP | |
Polyclonal | |
Unconjugated | |
CRTAP | |
cartilage associated protein, CASP, CRTAP, LEPREL3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10491, 363158, 56693 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75718, Q9CYD3 | |
Crtap | |
CRTAP Fusion Protein Ag8459 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |