missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CRTAP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | CRTAP |
---|---|
Concentration | 0.28 mg/mL |
Applications | Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835034
|
Proteintech
15724-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825034
|
Proteintech
15724-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The function of this protein remains unknown.Specifications
CRTAP | |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Rat, Human, Mouse | |
O75718, Q9CYD3 | |
10491, 363158, 56693 | |
CRTAP Fusion Protein Ag8459 | |
Primary | |
-20°C | |
CRTAP |
0.28 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
cartilage associated protein, CASP, CRTAP, LEPREL3 | |
Crtap | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |